Learn More
Medchemexpress LLC Neomangiferin | 64809-67-2 | 584.48 | 20 MG
SDP
Catalog No. 5000417054
Encompass_Preferred
Neomangiferin is an orally active natural flavonoid known for its multifaceted biological activities, including amelioration of non-alcoholic fatty liver disease (NAFLD), anti-colitis effects, and anti-aging properties. It also shows promise in preventing aseptic loosening of prostheses.
  • Partially ameliorates non-alcoholic fatty liver disease (NAFLD) by regulating gene expression related to free fatty acid uptake and lipid oxidation.
  • Exhibits anti-colitis effects by inhibiting Th17/Treg cell differentiation.
  • Exerts anti-aging and lifespan-extending effects by upregulating bas-1, which activates autophagy, IIS, and MAPK pathways.
  • May prevent aseptic loosening of prostheses after total joint arthroplasty due to anti-inflammatory and osteoclastogenesis-inhibiting properties.
Catalog No. 50-004-17054 Supplier Medchemexpress LLC Supplier No. HYN072320MG
Add to Cart
Add to Cart
Neomangiferin is an orally active natural flavonoid known for its multifaceted biological activities, including amelioration of non-alcoholic fatty liver disease (NAFLD), anti-colitis effects, and anti-aging properties. It also shows promise in preventing aseptic loosening of prostheses.
  • Partially ameliorates non-alcoholic fatty liver disease (NAFLD) by regulating gene expression related to free fatty acid uptake and lipid oxidation.
  • Exhibits anti-colitis effects by inhibiting Th17/Treg cell differentiation.
  • Exerts anti-aging and lifespan-extending effects by upregulating bas-1, which activates autophagy, IIS, and MAPK pathways.
  • May prevent aseptic loosening of prostheses after total joint arthroplasty due to anti-inflammatory and osteoclastogenesis-inhibiting properties.