Learn More
LABSYSTEMS DIAGNOSTICS OY CAH Multilevel Calibrator in DBS format
Catalog No. NC2170418
Encompass
NeoMass CAH Multilevel DBS Calibrators are used for the calibration of the analysis-system to determine the unknown concentration of analytes (Androstenedione, Cortisol, 11-Deoxycortisol, 21-Deoxycortisol, 17-Hydroxyprogesterone and 21-Hydroxyprogesterone) in Dried Blood Spots (DBS). These DBS calibrators are prepared by using human blood and are available as a calibrator set with 8 levels, including a blank. The calibrators are handled and used in the same way as newborn DBS samples.
This item is not returnable. View return policy
Catalog No. NC2170418 Supplier LABSYSTEMS DIAGNOSTICS OY Supplier No. 3467700
NeoMass CAH Multilevel DBS Calibrators are used for the calibration of the analysis-system to determine the unknown concentration of analytes (Androstenedione, Cortisol, 11-Deoxycortisol, 21-Deoxycortisol, 17-Hydroxyprogesterone and 21-Hydroxyprogesterone) in Dried Blood Spots (DBS). These DBS calibrators are prepared by using human blood and are available as a calibrator set with 8 levels, including a blank. The calibrators are handled and used in the same way as newborn DBS samples.