Learn More
Med Vet International Neomycin Antibacterial Liquid, 16oz, Neomycin Sulfate
Catalog No. 502828783
Encompass_Preferred
Used for treatment and control of colibacillosis (bacterial enteritis) caused by Escherichia coli susceptible to neomycin sulfate in cattle (excluding veal calves), swine, sheep and goats. May be given undiluted or diluted with water. Administer by drench or by adding to drinking water.
Catalog No. 50-282-8783 Supplier Med Vet International Supplier No. RXVNEOLIQUID
Used for treatment and control of colibacillosis (bacterial enteritis) caused by Escherichia coli susceptible to neomycin sulfate in cattle (excluding veal calves), swine, sheep and goats. May be given undiluted or diluted with water. Administer by drench or by adding to drinking water.