Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Neomycin C (hexaacetate) | 66-86-4 | 97.1% | 1 MG
SDP

Catalog No. 5000340723
Encompass_Preferred

Neomycin C hexaacetate is an impurity of Neomycin and a stereoisomer of Neomycin B. It exhibits in vitro antimicrobial activity against Staphylococcus epidermidis and Staphylococcus aureus, and can be used for the research of bacterial infection.

  • Exhibits in vitro antimicrobial activity against Staphylococcus epidermidis and Staphylococcus aureus.
  • Useful for bacterial infection research.
  • Molecular weight of 974.96.
  • Chemical formula C35H70N6O25.
  • Solid appearance.
  • White to off-white color.

Catalog No. 50-003-40723 Supplier Medchemexpress LLC Supplier No. HY131130A1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neomycin C hexaacetate is an impurity of Neomycin and a stereoisomer of Neomycin B. It exhibits in vitro antimicrobial activity against Staphylococcus epidermidis and Staphylococcus aureus, and can be used for the research of bacterial infection.

  • Exhibits in vitro antimicrobial activity against Staphylococcus epidermidis and Staphylococcus aureus.
  • Useful for bacterial infection research.
  • Molecular weight of 974.96.
  • Chemical formula C35H70N6O25.
  • Solid appearance.
  • White to off-white color.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.