Learn More
Medchemexpress LLC Neomycin sulfate | 1405-10-3 | 99.86% | 5 G
SDP
Catalog No. 5000408391
Encompass_Preferred
Neomycin sulfate is an aminoglycoside antibiotic that exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit, thereby blocking bacterial protein synthesis. It is also a known phospholipase C (PLC) inhibitor. This compound can be used for cancer research and is involved in various clinical trials for conditions such as Crohn's Disease and colorectal neoplasms.
  • Inhibits nuclear translocation of angiogenin
  • Abolishes angiogenin-induced proliferation of HUVE cells
  • Inhibits IP3-mediated Ca2+ release
  • Depletes gut microbiota in specific mouse models
  • Can be used for cancer research
Catalog No. 50-004-08391 Supplier Medchemexpress LLC Supplier No. HYB04705G
Neomycin sulfate is an aminoglycoside antibiotic that exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit, thereby blocking bacterial protein synthesis. It is also a known phospholipase C (PLC) inhibitor. This compound can be used for cancer research and is involved in various clinical trials for conditions such as Crohn's Disease and colorectal neoplasms.
  • Inhibits nuclear translocation of angiogenin
  • Abolishes angiogenin-induced proliferation of HUVE cells
  • Inhibits IP3-mediated Ca2+ release
  • Depletes gut microbiota in specific mouse models
  • Can be used for cancer research