Learn More
Medchemexpress LLC Neomycin (sulfate) | 1405-10-3 | 99.9% | 500 MG
SDP
Catalog No. 5000407636
Encompass_Preferred
Neomycin sulfate is an aminoglycoside antibiotic that exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit. It is used in various research applications, including as a phospholipase C inhibitor and angiogenesis modulator. It inhibits nuclear translocation of angiogenin, as well as angiogenin-induced cell proliferation and angiogenesis.
  • Aminoglycoside antibiotic
  • Inhibits nuclear translocation of angiogenin
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • For research use only
  • Recommended storage at 4°C, sealed, away from light and moisture. Avoid inhalation, contact with eyes and skin, and ensure adequate ventilation.
Catalog No. 50-004-07636 Supplier Medchemexpress LLC Supplier No. HYB0470500MG
Neomycin sulfate is an aminoglycoside antibiotic that exerts antibacterial activity through irreversible binding of the nuclear 30S ribosomal subunit. It is used in various research applications, including as a phospholipase C inhibitor and angiogenesis modulator. It inhibits nuclear translocation of angiogenin, as well as angiogenin-induced cell proliferation and angiogenesis.
  • Aminoglycoside antibiotic
  • Inhibits nuclear translocation of angiogenin
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • For research use only
  • Recommended storage at 4°C, sealed, away from light and moisture. Avoid inhalation, contact with eyes and skin, and ensure adequate ventilation.