Learn More
Medchemexpress LLC Neomycin sulfate | 1405-10-3 | 99.9% | 908.88 | C23H48N6O17S | 10 MG
SDP
Catalog No. 5000298065
Encompass_Preferred
Neomycin sulfate is an aminoglycoside antibiotic supplied as the sulfate salt and intended for research and analytical use. It binds irreversibly to the 30S ribosomal subunit to block bacterial protein synthesis and has reported activity as a phospholipase C inhibitor and inhibitor of angiogenin nuclear translocation.
  • Analytical standard for research and analytical applications.
  • High purity (approx. 99.9%).
  • Sulfate salt, solid form suitable for standard preparation.
  • Molecular weight 908.88 g/mol.
  • Packaged in 10 mg quantities for small-scale assays.
  • Suitable as a reference material for microbiology and biochemical assays.
Catalog No. 50-002-98065 Supplier Medchemexpress LLC Supplier No. HYB0470R10MG
Neomycin sulfate is an aminoglycoside antibiotic supplied as the sulfate salt and intended for research and analytical use. It binds irreversibly to the 30S ribosomal subunit to block bacterial protein synthesis and has reported activity as a phospholipase C inhibitor and inhibitor of angiogenin nuclear translocation.
  • Analytical standard for research and analytical applications.
  • High purity (approx. 99.9%).
  • Sulfate salt, solid form suitable for standard preparation.
  • Molecular weight 908.88 g/mol.
  • Packaged in 10 mg quantities for small-scale assays.
  • Suitable as a reference material for microbiology and biochemical assays.