Learn More
U.S. Pharmacopeia Neomycin Sulfate, 1405-10-3, MFCD00037007, 200mg
Catalog No. NC9894533
Encompass
Molecular formula C23H46N6O13 . 3H2SO4, Molecular weight 908.89, Synonyms: Neomycin B sulfate
Catalog No. NC9894533 Supplier U.S. Pharmacopeia Supplier No. 1458009
Molecular formula C23H46N6O13 . 3H2SO4, Molecular weight 908.89, Synonyms: Neomycin B sulfate