Learn More
Research Products International Corp Neomycin Sulfate, 500 G
SDP
Catalog No. 501977160
Encompass_Preferred
Neomycin Sulfate is an antibiotic that is derived from the bacterium Streptomyces fradiae and belongs to the aminoglycoside class of antibiotics. It exhibits broad-spectrum activity against Gram-negative and some Gram-positive bacteria. 
In molecular biology, Neomycin Sulfate is commonly used as a selective agent in genetic engineering experiments and cell culture applications. It is often included in growth media or buffers to prevent the growth of bacteria and ensure the purity of cell lines.
Neomycin Sulfate functions by inhibiting protein synthesis in bacteria, leading to cell death.  Neomycin Sulfate is also employed in microbiology research.
WARNING! This product can expose you to Neomycin sulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-197-7160 Supplier Research Products International Corp Supplier No. N20040500.0
Neomycin Sulfate is an antibiotic that is derived from the bacterium Streptomyces fradiae and belongs to the aminoglycoside class of antibiotics. It exhibits broad-spectrum activity against Gram-negative and some Gram-positive bacteria. 
In molecular biology, Neomycin Sulfate is commonly used as a selective agent in genetic engineering experiments and cell culture applications. It is often included in growth media or buffers to prevent the growth of bacteria and ensure the purity of cell lines.
Neomycin Sulfate functions by inhibiting protein synthesis in bacteria, leading to cell death.  Neomycin Sulfate is also employed in microbiology research.
WARNING! This product can expose you to Neomycin sulfate, a chemical which is known to the State of California to cause developmental harm. For more information go to www.P65Warnings.ca.gov.