Learn More
Med Vet International Neomycin (Neomycin Sulfate) Antibacterial Liquid, 1 Gallon, Each
Catalog No. 502828463
Encompass_Preferred
Neomycin suflate 200 mg/mL. Indicated for the treatment and control of colibacillosis (bacterial enteritis) caused by Escherichia coli susceptible to neomycin sulfate in cattle (excluding veal calves), swine, sheep and goats. May be given undiluted or diluted with water. Administer by drench or by adding to drinking water.
Catalog No. 50-282-8463 Supplier Med Vet International Supplier No. RXNEOMYCIN200V
Neomycin suflate 200 mg/mL. Indicated for the treatment and control of colibacillosis (bacterial enteritis) caused by Escherichia coli susceptible to neomycin sulfate in cattle (excluding veal calves), swine, sheep and goats. May be given undiluted or diluted with water. Administer by drench or by adding to drinking water.