Learn More
Crescent Chemical Co Inc NEOMYCIN-SULFATE RESEARCH
SDP
Supplier:  Crescent Chemical Co Inc SE30250.01
Encompass_Preferred
Neomycin-sulfate research grade, Ph. Eur. Min. 680 U/mg. Aminoglycoside antibiotic complex. Inhibits protein biosynthesis by binding to the 30S subunit of bacterial ribosomes. Causes misreading of mRNA. Size - 10G Storage Conditions - +2 °C TO +8 °C
Catalog No. 50-247-915