Learn More
Crescent Chemical Co Inc NEOMYCIN-SULFATE RESEARCH
SDP
Catalog No. 50247917
Encompass_Preferred
Neomycin-sulfate research grade, Ph. Eur. Min. 680 U/mg. Aminoglycoside antibiotic complex. Inhibits protein biosynthesis by binding to the 30S subunit of bacterial ribosomes. Causes misreading of mRNA. Size - 100G Storage Conditions - +2 °C TO +8 °C /
Catalog No. 50-247-917 Supplier Crescent Chemical Co Inc Supplier No. SE30250.03
Neomycin-sulfate research grade, Ph. Eur. Min. 680 U/mg. Aminoglycoside antibiotic complex. Inhibits protein biosynthesis by binding to the 30S subunit of bacterial ribosomes. Causes misreading of mRNA. Size - 100G Storage Conditions - +2 °C TO +8 °C /