Learn More
Medchemexpress LLC Neomycin (sulfate) (Standard) | 1405-10-3 | 99.87% | 100 MG
SDP
Catalog No. 5000408208
Encompass_Preferred
Neomycin sulfate (Standard) is the analytical standard of Neomycin sulfate. It is an aminoglycoside antibiotic that works by irreversibly binding to the nuclear 30S ribosomal subunit, blocking bacterial protein synthesis. It also acts as a phospholipase C (PLC) inhibitor and strongly inhibits the nuclear translocation of angiogenin, as well as angiogenin-induced cell proliferation and angiogenesis.
  • Intended for research and analytical applications
  • Exhibits antibacterial activity
  • Known as a phospholipase C (PLC) inhibitor
  • Inhibits nuclear translocation of angiogenin
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-004-08208 Supplier Medchemexpress LLC Supplier No. HYB0470R100MG
Neomycin sulfate (Standard) is the analytical standard of Neomycin sulfate. It is an aminoglycoside antibiotic that works by irreversibly binding to the nuclear 30S ribosomal subunit, blocking bacterial protein synthesis. It also acts as a phospholipase C (PLC) inhibitor and strongly inhibits the nuclear translocation of angiogenin, as well as angiogenin-induced cell proliferation and angiogenesis.
  • Intended for research and analytical applications
  • Exhibits antibacterial activity
  • Known as a phospholipase C (PLC) inhibitor
  • Inhibits nuclear translocation of angiogenin
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS