Learn More
Medchemexpress LLC Neomycin (sulfate) (Standard) | 1405-10-3 | 99.9% | 25 MG
SDP
Catalog No. 5000450595
Encompass_Preferred
Neomycin (sulfate) (Standard) is an analytical standard used for research and analytical applications. This aminoglycoside antibiotic works by inhibiting bacterial protein synthesis through irreversible binding to the nuclear 30S ribosomal subunit. It is also recognized as a phospholipase C (PLC) inhibitor and can potently inhibit nuclear translocation of angiogenin, as well as angiogenin-induced cell proliferation and angiogenesis.
  • Aminoglycoside antibiotic
  • Inhibits bacterial protein synthesis
  • Known phospholipase C (PLC) inhibitor
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • Used as an analytical standard in HPLC, GC, and MS experiments
Catalog No. 50-004-50595 Supplier Medchemexpress LLC Supplier No. 25MGHYB0470R
Neomycin (sulfate) (Standard) is an analytical standard used for research and analytical applications. This aminoglycoside antibiotic works by inhibiting bacterial protein synthesis through irreversible binding to the nuclear 30S ribosomal subunit. It is also recognized as a phospholipase C (PLC) inhibitor and can potently inhibit nuclear translocation of angiogenin, as well as angiogenin-induced cell proliferation and angiogenesis.
  • Aminoglycoside antibiotic
  • Inhibits bacterial protein synthesis
  • Known phospholipase C (PLC) inhibitor
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • Used as an analytical standard in HPLC, GC, and MS experiments