Learn More
Medchemexpress LLC Neomycin sulfate (Standard) | 1405-10-3 | 99.9% | 250 MG
SDP
Catalog No. 5000406516
Encompass_Preferred
Neomycin sulfate (Standard) is an analytical standard for research applications. This aminoglycoside antibiotic functions by irreversibly binding to the bacterial 30S ribosomal subunit, blocking protein synthesis. It also acts as a phospholipase C (PLC) inhibitor and inhibits nuclear translocation of angiogenin, along with angiogenin-induced cell proliferation and angiogenesis.
  • Analytical standard for research and analytical applications
  • Aminoglycoside antibiotic
  • Inhibits bacterial protein synthesis by binding to 30S ribosomal subunit
  • Functions as a phospholipase C (PLC) inhibitor
  • Potently inhibits nuclear translocation of angiogenin
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)
Catalog No. 50-004-06516 Supplier Medchemexpress LLC Supplier No. 250MGHYB0470R
Neomycin sulfate (Standard) is an analytical standard for research applications. This aminoglycoside antibiotic functions by irreversibly binding to the bacterial 30S ribosomal subunit, blocking protein synthesis. It also acts as a phospholipase C (PLC) inhibitor and inhibits nuclear translocation of angiogenin, along with angiogenin-induced cell proliferation and angiogenesis.
  • Analytical standard for research and analytical applications
  • Aminoglycoside antibiotic
  • Inhibits bacterial protein synthesis by binding to 30S ribosomal subunit
  • Functions as a phospholipase C (PLC) inhibitor
  • Potently inhibits nuclear translocation of angiogenin
  • Inhibits angiogenin-induced cell proliferation and angiogenesis
  • Used in qualitative, quantitative, and methodological research experiments (HPLC, GC, MS)