Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dukal™ Neon Adhesive Bandages

Catalog No. 22415759
Change view
Click to view available options
Dimensions (L x W):
0.75 x 3 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
22-415-759 0.75 x 3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-415-759 Supplier Dukal™ Supplier No. 1076413
Add to Cart
Add to Cart
This item is not returnable. View return policy

These Neon Adhesive Bandages come in three colors.

  • Available in yellow, orange and pink.
  • Help kids forget their pain with these fun designs.
  • Comes with the preferred and patented Stat Strip™ easy opening wrapper that nurses request.
  • Features a highly absorbent, non-adherent wound pad.

Specifications

Material Plastic
Product Type Adhesive Bandage
Sterility Non-sterile
Dimensions (L x W) 0.75 x 3 in.
For Use With (Application) For minor cuts and scrapes
Color Neon Yellow, Orange and Pink
Latex Free Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.