Learn More
GENERAL BIOPHYSICS LLC Neon Gas Analyzer Model A
Catalog No. NC3361684
Encompass
Thermoconductivity Gas Analyzer for Neon with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.4%.
Catalog No. NC3361684 Supplier GENERAL BIOPHYSICS LLC Supplier No. GANEO2A
Add to Cart
Add to Cart
Request a Quote
Thermoconductivity Gas Analyzer for Neon with Onboard PumpApplication: Calibrated for argon gas with onboard pump to take gas samples.Features: Includes data display screen.Accuracy: ±0.4%.