Learn More
GENERAL BIOPHYSICS LLC Neon Gas Analyzer Model B
Catalog No. NC3361685
Encompass
Thermoconductivity Gas Analyzer for Neon (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.
Catalog No. NC3361685 Supplier GENERAL BIOPHYSICS LLC Supplier No. GANEO2B
Thermoconductivity Gas Analyzer for Neon (In-line Monitoring)Application: Real-time, in-line concentration monitoring for xenon gas.Accuracy: ±1%.