Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Pipette
Catalog No. NEON8P
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 pipette
Catalog No. Quantity
NEON8P 1 pipette
1 options
Catalog No. NEON8P Supplier Invitrogen™ Supplier No. NEON8P
Add to Cart
Add to Cart
The Neon NxT Electroporation System 8-Channel Pipette is designed specifically for use with the Neon NxT Electroporation System.
The Neon NxT Electroporation System 8-Channel Pipette is designed specifically for use with the Neon NxT Electroporation System (Cat. No. NEON18S). Each Neon NxT Electroporation System with 1-Channel and 8-Channel Pipettes (Cat. Nos. NEON18S and NEON18SK) comes with an 8-channel pipette. Those looking to upgrade their Neon NxT Electroporation System with the 1-Channel Pipette (Cat. Nos. NEON1S, NEON1SK) should pursue the Neon NxT Electroporation System 8-Channel Upgrade Package (Cat. Nos. NEON8 and NEON8U), which includes the Neon NxT Electroporation System 8-Channel Pipette, 8-channel pipette station, and 8-channel software activation. This pipette is offered separately for those needing an additional 8-channel pipette or replacement for their existing pipette.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Transfection
For Use With (Equipment) Neon™ NxT Electroporation System
Quantity 1 pipette
Tip Type Neon™ NxT 10-μL and 100-μL
For Research Use Only. Not for use in diagnostic procedures.