Learn More
Invitrogen™ Neon™ NxT Electroporation System 8-Channel Upgrade Package
Catalog No. NEON8
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 package
Catalog No. Quantity
NEON8 1 package
1 options
Catalog No. NEON8 Supplier Invitrogen™ Supplier No. NEON8
Add to Cart
Add to Cart
Request a Quote
The Neon NxT Electroporation System 8-Channel Upgrade Package is available for the Neon NxT Electroporation System with 1-Channel Pipette.
The Neon NxT Electroporation System 8-Channel Upgrade Package is available for the Neon NxT Electroporation System with 1-Channel Pipette (Cat. Nos. NEON1S and NEON1SK). The upgrade package includes the Neon NxT 8-channel pipette, 8-channel pipette station, 8-channel software activation, and a 1-year warranty that covers the device, both pipettes, and both stations.
The Neon NxT Electroporation System 8-Channel Pipette enables the electroporation of 96 samples in less than 15-minutes while allowing for full flexibility over each reaction. Due to the 8-channel tube design, each tip is separated in its own chamber and isolated from the other tips, allowing for different electroporation parameters, resuspension buffer, delivery payload, or even cell type. Additionally, the Neon NxT 8-channel pipette utilizes the same Neon NxT tips and buffers as the 1-channel pipette.
TRUSTED_SUSTAINABILITY
Specifications
Compatible Tips Neon™ NxT
Number of Channels 8
Volume (Metric) 10 μL, 100 μL
For Use With (Application) Transfection
For Use With (Equipment) Neon™ NxT Electroporation System (Cat. No. NEON1S, NEON1SK)
Includes Neon™ NxT 8-channel pipette, 8-channel pipette station, 8-channel software activation, and quick reference guide
Quantity 1 package
Warranty 1-year Standard Warranty (Covers: Neon™ NxT electroporation device, 1-channel pipette, 8-channel pipette, 1-channel pipette station, 8-channel pipette station)
For Research Use Only. Not for use in diagnostic procedures.