Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Neon™ Transfection System

Catalog No. mpk5000
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MPK5000 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MPK5000 Supplier Invitrogen™ Supplier No. MPK5000

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Includes

Neon electroporation device, pipette, pipette station, USB thumbdrive, manual and power cord.

For stem cells, primary cells, hard-to-transfect cells and established cell lines

Enables fast and efficient delivery of nucleic acids into all mammalian cell types, including primary, stem, and difficult-to-transfect cells. The flexible and open system allows the user to perform high-quality transfections using optimized or user-defined protocols in three simple steps with as few as 2 × 104 cells per reaction and a reaction time of just 1 second. A novel reaction chamber provides a dramatic increase in transfection efficiency and cell viability.

  • Up to 90% in many cell types, including difficult-to-transfect cells, primary, and stem cells
  • Easily transfect from 2 × 104 cells to 6 × 106 cells per reaction
  • Easy to use, with a single reagent kit for all cell types
  • Open system allows electroporation parameters to be optimized freely
  • One-of-a-kind electroporation
  • Plug-n-play out of the box
  • Perform more transfections with limited or precious cells

For use at room temperature under normal laboratory conditions.

Adenoviral shRNA, Cell Culture, Lentiviral shRNA & miRNA, Plasmid Transfection, RNAi, RNAi Transfection, RNAi, Epigenetics & Noncoding RNA Research, Retroviral RNAi, Stem Cell & Primary Cell Transfections, Synthetic siRNA Transfection, Transfection, Vector-Based RNAi, siRNA

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY

Specifications

Amperage 3.0-6.0 A
Classification Animal Origin-Free, Chemically Defined, Low-Protein, Xeno-Free, Reduced Growth Factors, Reduced Serum
Content And Storage The Neon™ Transfection System comes with a Neon™ electroporation device, pipette, pipette station, USB thumbdrive, manual and power cord.

For use at room temperature under normal laboratory conditions.
Cell Type Stem Cells, Primary Cells, Hard-to-Transfect Cells
Format 6-well Plate, 12-well Plate, 24-well Plate, 48-well Plate, 96-well Plate, Flasks
Sample Type Plasmid DNA, Synthetic siRNA
For Use With (Equipment) Neon™ Transfection System
For Use With (Application) Transfection
Max. Voltage 0.5-2.5 kV
Product Line Neon
Product Type Transfection System
Purity Transfection Grade
Quantity 1 Ea.
Reaction Time 1 sec
Serum Compatible Yes
Shipping Condition Room Temperature
Starting Material Amount 2x10^4 - 6 x10^8 cells; Varies by cell type
System Type Neon™ Transfection System
Voltage 100-240 VAC
Wattage 300 W
Volume (Metric) Sample Depends on kit used
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.