Learn More
Worthington Biochemical Corporation NEONATAL CARDIO-MYOCYTE ISO/KT
SDP
Catalog No. NC9988985
Encompass
Kit for performing five separate tissue dissociations each containing up to twelve hearts. Contains single use vials of purified collagenase and trypsin, CMF-HBSS, Leibovitz L-15 media and Falcon cell strainers along with a detailed protocol.
Catalog No. NC9988985 Supplier Worthington Biochemical Corporation Supplier No. LK003300
Kit for performing five separate tissue dissociations each containing up to twelve hearts. Contains single use vials of purified collagenase and trypsin, CMF-HBSS, Leibovitz L-15 media and Falcon cell strainers along with a detailed protocol.