Learn More
Poltex Neonatal Universal Circuit Holder
SDP
Catalog No. 22000336
Change view
Click to view available options
Dimensions (L x W x H):
6 x 6 x 8 to 10 in.
Catalog No. Dimensions (L x W x H)
22-000-336 6 x 6 x 8 to 10 in.
1 options
Catalog No. 22-000-336 Supplier Poltex Supplier No. NUCH
Get a Poltex Pipette Holder with $3,000 Purchase
View all promotions
Add to Cart
Add to Cart
Neonatal Universal Circuit Holder aids in securing oscillator (HFOV), ventilator and IV tubing in the Neonatal Intensive Care Unit (NICU) isolette or warmer
  • Wheel rotates and/or slides up and down in a slot for easy adjustment
  • Wheel holds firmly in place with cam lever
  • Circuit Holder is secured under the mattress on any side of an open warmer or incubator, and can be easily moved to accommodate any procedure.
Specifications
Autoclavable Not Autoclavable
Dimensions (L x W x H) 6 x 6 x 8 to 10 in.