Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentyl chloride, 98%

Catalog No. AAL0236506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0236506 5 g
AAL0236514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0236506 Supplier Thermo Scientific Chemicals Supplier No. L0236506
Add to Cart

CAS: 753-89-9 | C5H11Cl | 106.593 g/mol

Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Neopentyl chloride is a substrate used for quantum mechanics calculations on LinB to study about active site conformations and protonation states for catalysis.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and alkali metals.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-89-9
Molecular Formula C5H11Cl
Molecular Weight (g/mol) 106.593
MDL Number MFCD00000941
InChI Key JEKYMVBQWWZVHO-UHFFFAOYSA-N
PubChem CID 12956
IUPAC Name 1-chloro-2,2-dimethylpropane
SMILES CC(C)(C)CCl

Specifications

Melting Point -20°C
Density 0.868
Boiling Point 83°C to 84°C
Flash Point −8°C (17°F)
Odor Sweet
Refractive Index 1.4039
Linear Formula (CH3)3CCH2Cl
Quantity 5 g
UN Number UN1107
Beilstein 1730985
Synonym neopentyl chloride, propane, 1-chloro-2,2-dimethyl, neo-pentylchloride, t-c4h9ch2cl, 2,2-dimethylpropyl chloride, acmc-1bc1q, neopentyl chloride 5g, propane,1-chloro-2,2-dimethyl, 1-chloro-2,2-dimethylpropane
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 106.6
Percent Purity 98%
Chemical Name or Material Neopentyl chloride
Show More Show Less
missing translation for 'healthHazard1' H225
missing translation for 'healthHazard2' GHS H Statement
H225
Highly flammable liquid and vapor.
missing translation for 'healthHazard3' P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMYL CHLORIDES
EINECSNumber 212-040-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.