Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Neopentyl glycol diacrylate

Catalog No. 408255100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-825-5100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-825-5100ML Supplier Sigma Aldrich Supplier No. 408255100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Neopentyl glycol diacrylate

Specifications

CAS 2223-82-7
Density 1.031 g/mL (at 25°C)
Molecular Formula C11H16O4
Linear Formula (H2C=CHCO2CH2)2 C(CH3)2
Refractive Index n20/D 1.453 (literature)
MDL Number MFCD00048148
Quantity 100 mL
Synonym (2,2-Dimethyl-3-prop-2-enoyloxypropyl) prop-2-enoate; 2,2-Dimethylpropanediol diacrylate; 2-Propenoic acid, 2,2-dimethyl-1,3-propanediyl ester; Neopentanediol diacrylate
Molecular Weight (g/mol) 212.24
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.