Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Neopentyl glycol diglycidyl ether

Catalog No. 338036100ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-803-6100ML 100 mL
33-803-6500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-803-6100ML Supplier Sigma Aldrich Supplier No. 338036100ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Neopentyl glycol diglycidyl ether (NGDE) is a low toxic epoxy molecule with two epoxy pendant groups at the end of the long chain. It is majorly used as a reactive diluent for epoxy resin. It facilitates the resin with high strength and impact resistance.NGDE can be used in the synthesis of epoxy polymers for the development of shape memory polymers. It can also be used in the photopolymerization of cationic polymers.

Specifications

CAS 17557-23-2
Density 1.04 g/mL (at 25°C)
Boiling Point 103°C to 107°C (1 mmHg)
Molecular Formula C11H20O4
Refractive Index n20/D 1.457 (literature)
Linear Formula C11H20O4
Quantity 100 mL
MDL Number MFCD00040687
Synonym 1,3-Bis(2,3-epoxypropoxy)-2,2-dimethylpropane; 2,2 -[(2,2-Dimethyl-1,3-propanediyl)bis(oxymethylene)]bis[oxirane]
Molecular Weight (g/mol) 216.27
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.