Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neopentylamine, 97%

Catalog No. AC173691000
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC173691000 100 mL
AC173690250 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC173691000 Supplier Thermo Scientific Chemicals Supplier No. 173691000
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 5813-64-9 | C5H13N | 87.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5813-64-9
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00008134
InChI Key XDIAMRVROCPPBK-UHFFFAOYSA-N
Synonym neopentylamine, 2,2-dimethylpropylamine, 1-propanamine, 2,2-dimethyl, 2,2-dimethyl-1-propylamine, 1-amino-2,2-dimethylpropane, neo-c5h11nh2, 2,2-dimethyl-1-propylamin, neo-pentylamine, neopentyl amine, 2,2-dimethylpropanamine
PubChem CID 79882
IUPAC Name 2,2-dimethylpropan-1-amine
SMILES CC(C)(C)CN

Specifications

Density 0.7400g/mL
Boiling Point 79.0°C to 82.0°C
Flash Point −13°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Refractive Index 1.403
Linear Formula (CH3)3CCH2NH2
Quantity 100 mL
Beilstein 04,188
Specific Gravity 0.74
Solubility Information Solubility in water: soluble.
Formula Weight 87.16
Percent Purity 97%
Chemical Name or Material Neopentylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective g
EINECSNumber 227-378-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.