Learn More
DIAGNOSTICA STAGO INC STA-Neoplastine CI Plus 5, 6 x 5 mL
Catalog No. NC0591432
Encompass
Determination of Prothrombin Time (PT) by STA analyzers (ISI~1.3). Freeze-dried rabbit brain thromboplastin with heparin inhibitor.
Catalog No. NC0591432 Supplier DIAGNOSTICA STAGO INC Supplier No. 00606
Determination of Prothrombin Time (PT) by STA analyzers (ISI~1.3). Freeze-dried rabbit brain thromboplastin with heparin inhibitor.