Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Perfex™ Neoprene Quick-Dry Foam Squeegees

Catalog No. 17998618 Shop All Perfex Corp. Products
Change view
Click to view available options
Width (English):
18 in.
24 in.
Color:
Black
Blue
Green
Orange
Purple
Red
White
Yellow
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English) Color
17-998-618 24 in. Yellow
17-998-604 18 in. Blue
17-998-605 18 in. Black
17-998-606 18 in. Green
17-998-607 18 in. Orange
17-998-608 18 in. Purple
17-998-609 18 in. Red
17-998-610 18 in. White
17-998-611 18 in. Yellow
17-998-612 24 in. Blue
17-998-613 24 in. Black
17-998-614 24 in. Green
17-998-615 24 in. Orange
17-998-616 24 in. Purple
17-998-617 24 in. Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
Catalog No. 17-998-618 Supplier Perfex™ Supplier No. 9724Y
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Neoprene foam floor squeegees with flexible blades, 18 or 24 in. widths, and color options.

  • Neoprene rubber foam within a durable polymer channel. Quickly wipes surfaces dry and nearly streak-free.
  • Flexible blades allow use on uneven or irregular surfaces, excellent for grouted floors and quarry tile.
  • Ideal for floor chemical application. Resistant to water, oil, grease, detergents, sanitizer, and solvents. Wash and rinse clean with warm water. Lightweight and non-marring.
  • Connects to the long variant of the Lite-N-Tite connectors only.
  • Available in 18 in. and 24 in. blade widths. Eight color options are available to color-code equipment per facility zones. Including White, Red, Blue, Green, Yellow, Orange, Purple, and Black.

Specifications

Product Type Floor Squeegee
Color-coded Yes
Color Yellow
Material Neoprene Rubber Foam
Width (English) 24 in.
Width (Metric) 60.96 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.