Learn More
Research Products International Corp Laboratory Grade Rubber Stoppers, Green Neoprene Rubber, Size #8, 41 x 33 x 25mm, 12 per Package
SDP
Catalog No. 50998958
Encompass
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available in green neoprene rubber that is resistant to chemicals and ozone deterioration or standard black SBR rubber. Solid rubber stoppers are molded in the USA according to standard industry size specification. Packaged in approximately one pound packages.
Catalog No. 50-998-958 Supplier Research Products International Corp Supplier No. 2591008
Laboratory Grade Rubber Stoppers offered in a wide range of common sizes for most laboratory applications. Available in green neoprene rubber that is resistant to chemicals and ozone deterioration or standard black SBR rubber. Solid rubber stoppers are molded in the USA according to standard industry size specification. Packaged in approximately one pound packages.