Learn More
Medchemexpress LLC HY-W011053 500mg Medchemexpress, Neotame CAS:165450-17-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYW011053500MG
Encompass_Preferred
Medchemexpress, HY-W011053 500mg Neotame CAS:165450-17-9 Neotame is a derivative of Aspartame and is a low-caloric and high-intensity artificial sweetener that is 7000-13,000 times sweeter than sugar. Neotame is a non-nutritive sweetener and flavor enhancer that can be used in a variety of foods. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3306