Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neotame, ≥98.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013309 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3309 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3309 Supplier MilliporeSigma Supelco Supplier No. 49777100MG
Add to Cart
Add to Cart

Neotame is a white colored, dipeptide methyl ester, which is found to be a highly potent and non-nutritive sweetener or flavour enhancer and hence finds applications in variety of foods. It is manufactured from aspartame and 3,3-dimethylbutyraldehyde in a one-step chemical synthesis process.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 165450-17-9
Physical Form Neat
Linear Formula C20H30N2O5
MDL Number MFCD09039056
Synonym N-[N-(3,3-Dimethylbutyl)-L-alpha-aspartyl]-L-phenylalanine 1-methyl ester
Type Analytical Standard
Quantity 100 mg
Formula Weight 378.46
Percent Purity ≥98.0% (HPLC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.