Learn More
AdipoGen Neoxaline (1 mg)
SDP
Supplier:  AdipoGen AGCN20154M001
Encompass_Preferred
Chemical. CAS: 71812-10-7. Formula: C23H25N5O4. MW: 435.5. Isolated from Aspergillus japonicus Fg-551. Antimitotic. Cell cycle inhibitor at the G2/M phase. Antiproliferative. Weak CNS stimulant with weak inhibitory activity on blood platelet aggregation. Mycotoxin.
Catalog No. 50-205-3536