Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Neoxanthin, ≥97.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111013141 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3141 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3141 Supplier MilliporeSigma Supelco Supplier No. 729941MG
Add to Cart
Add to Cart

Neoxanthin is an allenic xanthophyll and a precursor of the plant hormone abscisic acid, which is formed as the last product of carotenoid synthesis in green plants.

Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 14660-91-4
Linear Formula C40H56O4
Synonym (3 S,3 ′S,5 R,5 ′R,6 R,6 ′S,9 ′-cis)-6,7-Didehydro-5 ′,6 ′-epoxy-5 ′,6 ′-dihydro-beta,beta-carotene-3,3 ′,5(6 H)-triol; 9 ′-cis-Neoxanthin
Type Analytical Standard
Quantity 1 mg
Formula Weight 600.87
Percent Purity ≥97.0% (HPLC)
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.