Learn More
TARGETMOL CHEMICALS INC NEP-IN-1 5MG
SDP
Catalog No. 502581617
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NEP-IN-1 is an inhibitor of neutral endopeptidase (NEP IC50 = 2 nM) and can be used in studies about female sexual arousal disorders. purity: 98%
Catalog No. 50-258-1617 Supplier TARGETMOL CHEMICALS INC Supplier No. T109185MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. NEP-IN-1 is an inhibitor of neutral endopeptidase (NEP IC50 = 2 nM) and can be used in studies about female sexual arousal disorders. purity: 98%