Learn More
Bulldog Bio Inc NEPA Porator Electroporator, 1 System
SDP
Supplier:  Bulldog Bio Inc NEPA01
Encompass_Preferred
Impedance checks will give you better resultsElectroporation is a process by which one or more electrical pulses are delivered to a biological sample. These pulses result in the temporary disruption of the cell membrane, and under some conditions the cell wall. The “porated” cells are then able to quickly take up DNA, RNA, or proteins in the surrounding buffer. The NEPA Porator is designed to deliver one or two DC exponential decay waves with constant voltage. What makes the NEPA Porator so special is its ability to first check the resistance of the sample via a trickle charge. This quick check ensures your samples’ buffering conditions are in the right range, so you know that you’re applying the correct amount of energy each and every time. In many cases, a quick adjustment to the sample volume in the cuvette can bring resistance back into the optimal range so that you can proceed with your experiment.
Catalog No. NC1933461
Add to Cart
Request a Quote