Learn More
Bellco Glass, Inc. Nephelo Flask-Screw Cap 38, 300 mL
SDP
Supplier:  Bellco Glass, Inc. 257412133
Encompass_Preferred
Ideal for growing, storing and mixing or aerobic and anaerobic cultures. Threaded neck and sidearm with clean-out port. Supplied with 38mm and 18mm rubber lined black phenolic screw caps.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 300mLIncludes: 38mm And 18mm Black Phenolic Screw CapsSidearm Dimensions: 12x130mmCase Quantity: 4
Catalog No. 50-194-5572