Learn More
Bellco Glass, Inc. Nephlo Flask-DeLong,125mL
SDP
Catalog No. 501945569
Encompass_Preferred
Nephelo culture flask is ideal for growing, storing and mixing of aerobic and anaerobic cultures. Supplied with 25mm Bellco stainless steel closures.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 125mlIncludes: Bellco Stainless Steel Closure (2005 Series)Sidearm Dimensions: 14x130mmCase Quantity: 4
Catalog No. 50-194-5569 Supplier Bellco Glass, Inc. Supplier No. 257114130
Nephelo culture flask is ideal for growing, storing and mixing of aerobic and anaerobic cultures. Supplied with 25mm Bellco stainless steel closures.. MADE IN THE USASpecifications:Type 1 Class A Borosilicate GlassSizes: 125mlIncludes: Bellco Stainless Steel Closure (2005 Series)Sidearm Dimensions: 14x130mmCase Quantity: 4