Learn More
Bellco Glass, Inc. Nephlo Flask-DeLong, 300mL
SDP
Catalog No. 501945570
Encompass_Preferred
Nephelo culture flask is ideal for growing, storing and mixing of aerobic and anaerobic cultures. Supplied with 25mm Bellco stainless steel closures.Specifications:Type 1 Class A Borosilicate GlassSizes: 300mlIncludes: Bellco Stainless Steel Closure (2005 Series)Sidearm Dimensions: 14x130mmCase Quantity: 4
Catalog No. 50-194-5570 Supplier Bellco Glass, Inc. Supplier No. 257114133
Nephelo culture flask is ideal for growing, storing and mixing of aerobic and anaerobic cultures. Supplied with 25mm Bellco stainless steel closures.Specifications:Type 1 Class A Borosilicate GlassSizes: 300mlIncludes: Bellco Stainless Steel Closure (2005 Series)Sidearm Dimensions: 14x130mmCase Quantity: 4