Learn More
Enzo Life Sciences Nephrin (Ig-like domain 5) clonal antibody (Y17-R) (100µl)
SDP
Supplier:  Enzo Life Sciences ALX810016R100
Encompass_Preferred
Clone: Y17-R. Isotype: IgG. Immunogen: Synthetic peptide corresponding to an epitope in the centre of the fifth Ig-like domain of human nephrin (between aa 465 and 528). Species reactivity: Human, Mouse, Rat. Applications: ELISA, IHC (PS), WB. Purity: Major clone of IgGs obtained from immunoaffinity purified immunoglobulins corresponding to immunogenic peptide. Formulation: Liquid. In 20mM TRIS/HCl, pH 8.0, containing 10mg/ml BSA and 0.05% sodium azide.
Catalog No. 50-201-0075