Learn More
Medchemexpress LLC HY-D0885B 100mg Medchemexpress, Phosphocreatine (disodium) CAS:922-32-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYD0885B100MG
Encompass_Preferred
Medchemexpress, HY-P7392 10ug Nephroblastoma Overexpressed Gene Protein (CHO-expressed), Human CAS: Recombinant Human Nephroblastoma Overexpressed Gene Protein (CHO-expressed), an angiogenic regulator, supports cell adhesion, induces directed cell migration (chemotaxis), and promotes cell survival. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0189