Learn More
Apexbio Technology LLC Nepicastat (SYN-117) HCl 170151-24-3 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062573
Encompass_Preferred
Nepicastat (SYN-117) HCl is a potent and selective inhibitor targeting dopamine -hydroxylase (DBH) an enzyme responsible for converting dopamine to norepinephrine in catecholamine biosynthesis pathways By inhibiting DBH activity Nepicastat decreases norepinephrine synthesis and concurrently elevates dopamine levels Preclinical studies conducted in animal models such as spontaneously hypertensive rats and canine models demonstrate a dose-dependent reduction of norepinephrine levels accompanied by elevation of dopamine concentration across central nervous system and cardiac tissues Nepicastat serves as a valuable research tool in investigating catecholamine homeostasis cardiovascular function sympathetic regulation and dopamine-mediated neurotransmission
Catalog No. 50-000-62573 Supplier Apexbio Technology LLC Supplier No. A43325.1
Nepicastat (SYN-117) HCl is a potent and selective inhibitor targeting dopamine -hydroxylase (DBH) an enzyme responsible for converting dopamine to norepinephrine in catecholamine biosynthesis pathways By inhibiting DBH activity Nepicastat decreases norepinephrine synthesis and concurrently elevates dopamine levels Preclinical studies conducted in animal models such as spontaneously hypertensive rats and canine models demonstrate a dose-dependent reduction of norepinephrine levels accompanied by elevation of dopamine concentration across central nervous system and cardiac tissues Nepicastat serves as a valuable research tool in investigating catecholamine homeostasis cardiovascular function sympathetic regulation and dopamine-mediated neurotransmission