Learn More
Medchemexpress LLC Nepinalone hydrochloride | 22443-55-6 | 99.6% | 307.86 g/mol | C18H26ClNO | 1 ML
SDP
Catalog No. 5000239970
Encompass_Preferred
Nepinalone hydrochloride is an alkylaminated β-tetralone derivative used in research as a non-opioid, orally active antitussive. Supplied as a high-purity solid or as a ready-to-use 10 mM solution in DMSO, it is suitable for in vitro studies and for preparation of in vivo dosing formulations.
Catalog No. 50-002-39970 Supplier Medchemexpress LLC Supplier No. HY106873A10MM 1ML
Nepinalone hydrochloride is an alkylaminated β-tetralone derivative used in research as a non-opioid, orally active antitussive. Supplied as a high-purity solid or as a ready-to-use 10 mM solution in DMSO, it is suitable for in vitro studies and for preparation of in vivo dosing formulations.