Learn More
Medchemexpress LLC Neplanocin A | 72877-50-0 | 99.8% | 1 MG
Supplier: Medchemexpress LLC HY1304301MG
Neplanocin A is an antitumor antibiotic that exhibits significant antitumor activity against murine L1210 leukemia. It is also an irreversible inhibitor of AdoHcy hydrolase (Ki=8.39 nM). Additionally, Neplanocin A possesses antiviral activity and is effective against vaccinia virus. It is obtained from Ampulariella regularis.
- Antitumor antibiotic
- Antiviral activity
- Irreversible inhibitor of AdoHcy hydrolase (Ki=8.39 nM)
- Obtained from Ampullariella regularis