Learn More
Cayman Chemical Nepodn 10mg
SDP
Catalog No. 502939809
Encompass_Preferred
ABlactamase inhibitor (Kis 29-110 nM for class A and class C enzymes) selective for bacterialBlactamases over a panel of mammalian serine proteases (IC50s 1000 M) potentiates the activity of biapenem against K pneumonia expressing KPC-2 at 0 02 g/ml potentiates the activity of carbapenem antibiotics against clinical isolates of various bacteria (MICs 0 06-4 and 4-64 g/ml in the presence and absence of compound respectively) reduces the number of CFUs in the lung in a neutropenic mouse lung infection model when used in combination with biapenem or meropenem at 50 mg/kg
Catalog No. 50-293-9809 Supplier Cayman Chemical Supplier No. 27611 10MG
ABlactamase inhibitor (Kis 29-110 nM for class A and class C enzymes) selective for bacterialBlactamases over a panel of mammalian serine proteases (IC50s 1000 M) potentiates the activity of biapenem against K pneumonia expressing KPC-2 at 0 02 g/ml potentiates the activity of carbapenem antibiotics against clinical isolates of various bacteria (MICs 0 06-4 and 4-64 g/ml in the presence and absence of compound respectively) reduces the number of CFUs in the lung in a neutropenic mouse lung infection model when used in combination with biapenem or meropenem at 50 mg/kg