Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human Neprilysin (MME) ELISA Kit

Catalog No. EHMME
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 x 96 Tests
96 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EHMME 96 Tests
EHMMEX5 5 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. EHMME Supplier Invitrogen™ Supplier No. EHMME
Add to Cart
Add to Cart

ELISA

The Human Neprilysin (MME) ELISA quantitates Hu MME in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu MME. Principle of the method The Human MME solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

CD10, also known as the Common Acute Lymphocytic Leukemia Antigen (CALLA) and neutral endopeptidase (NEP), is a Zn2+-dependent metallo-peptidase with neutral metalloendopeptidase activity. It is a 100 kDa type II transmembrane glycoprotein encoded by a gene that exists in a single copy of greater than 45 kb. The 5' untranslated region of the CD10 gene is alternatively spliced, resulting in four separate mRNA transcripts, although the coding region remains unaffected by alternative splicing. CD10 is involved in the regulation of chemotactic and inflammatory processes involving neutrophils and plays a role in stromal cell-dependent B lymphopoiesis. It is expressed on immature B cells in adult bone marrow, mature B cells in germinal centers, and cells from patients with chronic myelocytic leukemia (CML). Additionally, CD10 is present on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas. Beyond the hematopoietic compartment, CD10 is highly expressed on the brush border of enterocytes and renal tubules and glomeruli, as well as on breast myoepithelial cells, bile canaliculi, and fibroblasts. CD10 cleaves peptides at the amino side of hydrophobic residues, inactivating several biologically active peptides, including endothelin, glucagon, gastrin, neurotensin, bradykinin, enkephalins, substance P, oxytocin, and others. Diseases associated with CD10 dysfunction include spinocerebellar ataxia 43 and Charcot-Marie Tooth Disease, highlighting its significance in both normal physiological processes and disease states.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08473
Assay Range 0.41-100 ng/mL
Assay Sensitivity 0.4 ng/mL
Calibration Range 0.4 ng/mL to 100 ng/mL
Content And Storage Pre-coated plate(s), standard or calibrator, detector antibody, HRP conjugate, diluents, wash buffer, substrate(s), and stop solution. Store kit at 2-8°C. See product manual for detailed contents and storage conditions for maximum stability.
Assay Solid-phase sandwich ELISA
Conjugate HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Protein Family Cytokines & Receptors
Sample Type Plasma, Serum, Supernatant
Specificity This ELISA kit shows no cross-reactivity with the following cytokines tested: human ADAM-9, Angiopoietin 2, APRIL, BMP-2, BMP-9, C5a, Cathepsin L, CD200, CD97, Chemerin, DcR3, FABP2, FAP, FGF-19, Galectin-3, HGF R, IFN-gammalpha, IGF-2, IGF-2 R, IL-1R6, IL-24, IL-33, Kallikrein 14, Legumain, LOX-1, MBL, Notch-1, NOV, Osteoactivin, PD-1, PGRP-5, Serpin A4, sFRP-3, Thrombomodulin, TLR2, TRAIL R1, Transferrin, WIF-1 ACE-2, Albumin, AMICA, Angiopoietin 4, BAFF, CA19-9, CD163 , Clusterin, CRTAM, CXCL14, Cystatin C, Decorin , Dkk-3, DLL1, Fetuin A, aFGF, FOLR1, Furin, GASP-1, GASP-2, GCSF R, HAI-2, IL-17 B R, IL-27, LAG-3 , LDL R, Pepsinogen I, RANK, RBP4, SOST, Syndecan-1, TACI, TFPI, TSP-1, TRAIL R2 , TRANCE, Troponin I, uPA, VE-Cadherin, WISP-1
Sufficient For 96 assays
Target Species Human
For Use With (Application) ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias 6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase6030454K05Rik; Atriopeptidase; C85356; CALLA; CD10; CMT2T; common acute lymphoblastic leukemia antigen; common acute lymphocytic leukemia antigen; Enkephalinase; EPN; membrane metallo endopeptidase; membrane metalloendopeptidase; membrane metallo-endopeptidase; membrane metallo-endopeptidase (neutral endopeptidase, enkephalinase, CALLA, CD10); membrane metallo-endopeptidase (neutral endopeptidase/enkephalinase); membrane metallo-endopeptidase variant 1; membrane metallo-endopeptidase variant 2; MME; NEP; neprilysin; neprilysin-390; neprilysin-411; neutral endopeptidase 24.11; SFE; Skin fibroblast elastase; CALLA; CD10; CMT2T; NEP; SCA43; SFE
Gene ID (Entrez) 4311
Gene Symbol Mme
Interassay CV <12%
Intraassay CV <10%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantity 96 Tests
Regulatory Status RUO
Research Discipline Cell Cycle & Proliferation
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 100 μL
Storage Requirements Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C
Synonym CALLA, CD10, NEP, SFE
Target CD10
Test Time 1 hr 20 min
Total Assay Time 4 hr 45 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.