Learn More
ACROBiosystems ACROBiosystems Human Neprilysin / MME / CD10 Protein, Tag Free
Supplier:  ACROBiosystems MMEH521B50UG
Encompass_Preferred
Recombinant Protein;50UG;Human Neprilysin, Tag Free (MME-H521b) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession # NP_000893.2).;This protein carries no "tag". The protein has a calculated MW of 80.0 kDa. The protein migrates as 80-100 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-195-6417