Learn More
ACROBiosystems Recombinant Protein;Human Neprilysin / MME / CD10 Protein, Fc Tag;HEK293;1MG;MME,MME,CALLA,CD10,CD-10,DKFZp686O16152,MGC126681,MGC126707,NEP,SFE,Neprilysin
Catalog No. 501657479
Encompass_Preferred
Recombinant Protein;Human Neprilysin / MME / CD10 Protein, Fc Tag;1MG;Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession # NP_000893.2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 107.3 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7479 Supplier ACROBiosystems Supplier No. MMEH526A1MG
Recombinant Protein;Human Neprilysin / MME / CD10 Protein, Fc Tag;1MG;Human Neprilysin, Fc Tag (MME-H526a) is expressed from human 293 cells (HEK293). It contains AA Tyr 52 - Trp 750 (Accession # NP_000893.2).;This protein carries a human IgG1 Fc tag at the N-terminus. The protein has a calculated MW of 107.3 kDa. The protein migrates as 120-130 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.