Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Nepsilon-(2,4-Dinitrophenyl)-L-lysine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. D10231G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D10231G 1 g
D1023100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D10231G Supplier TCI America Supplier No. D10231G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 14401-10-6
Molecular Formula C12H16ClN4O6
Molecular Weight (g/mol) 347.73
MDL Number MFCD00036383
InChI Key FJDUWQCVTQNLPA-UHFFFAOYNA-N
Synonym Nepsilon-Dnp-L-lysine Hydrochloride, H-Lys(Dnp)-OH.HCl
PubChem CID 22872935
IUPAC Name 2-amino-6-[(2,4-dinitrophenyl)amino]hexanoic acid hydrochloridyl
SMILES [Cl].NC(CCCCNC1=CC=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Color Yellow
Quantity 1 g
Formula Weight 348.74
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Nepsilon-(2,4-Dinitrophenyl)-L-lysine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.