Learn More
Medchemexpress LLC Nepsilon-Acetyl-L-lysine | 692-04-6 | 99.97% | 188.22 | 10 G
SDP
Catalog No. 5000436590
Encompass_Preferred
Nepsilon-Acetyl-L-lysine, an endogenous metabolite, is an R-chain N-acetylated α amino acid.
  • Endogenous metabolite
  • R-chain N-acetylated α amino acid
  • Solid appearance, white to off-white color
  • Soluble in water and PBS
  • For research use only
Catalog No. 50-004-36590 Supplier Medchemexpress LLC Supplier No. HY11342610G
Nepsilon-Acetyl-L-lysine, an endogenous metabolite, is an R-chain N-acetylated α amino acid.
  • Endogenous metabolite
  • R-chain N-acetylated α amino acid
  • Solid appearance, white to off-white color
  • Soluble in water and PBS
  • For research use only